ENA295068128 Enamine

methyl 1-(1-benzylpiperidin-4-yl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD16547553
  • Cas Number : 1188478-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62456
Availability details for
Quantity
  • Availability
  • Add to Cart