ENA295068136 Enamine

5-acetyl-1-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD16547562
  • Cas Number : 1240527-97-2
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62621
Availability details for
Quantity
  • Availability
  • Add to Cart