ENA295068140 Enamine

5-bromo-1-[2-(diethylamino)ethyl]-2,3-dihydro-1H-indole-2,3-dione hydrochloride

  • MDL number MFCD16547567
  • Cas Number : 1240529-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62658
Availability details for
Quantity
  • Availability
  • Add to Cart