ENA295068142 Enamine

3-(piperidin-4-ylidene)-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD16547572
  • Cas Number : 1240529-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62722
Availability details for
Quantity
  • Availability
  • Add to Cart