ENA295068144 Enamine

1-(4-amino-2-methylphenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD16547574
  • Cas Number : 1240529-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62806
Availability details for
Quantity
  • Availability
  • Add to Cart