ENA295068147 Enamine

N-[(thiophen-3-yl)methyl]aniline hydrochloride

  • MDL number MFCD16547577
  • Cas Number : 1240529-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62813
Availability details for
Quantity
  • Availability
  • Add to Cart