ENA295068148 Enamine

2-methyl-1,2,3,4-tetrahydroquinoline-4-carboxamide hydrochloride

  • MDL number MFCD16547578
  • Cas Number : 1240528-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62823
Availability details for
Quantity
  • Availability
  • Add to Cart