ENA295068151 Enamine

2-(3,5-dimethyl-1,2-oxazol-4-yl)butanoic acid

  • MDL number MFCD16547582
  • Cas Number : 1240528-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62852
Availability details for
Quantity
  • Availability
  • Add to Cart