ENA295068152 Enamine

2-{1-[5-(trifluoromethyl)pyridin-2-yl]-1H-1,2,3-triazol-4-yl}ethan-1-ol

  • MDL number MFCD16547583
  • Cas Number : 1240526-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63757
Availability details for
Quantity
  • Availability
  • Add to Cart