ENA295068156 Enamine

2-amino-N-methyl-N-[(pyridin-3-yl)methyl]acetamide dihydrochloride

  • MDL number MFCD16547587
  • Cas Number : 1240528-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63874
Availability details for
Quantity
  • Availability
  • Add to Cart