ENA295068157 Enamine

4-(hydrazinylmethyl)-2-methylquinoline trihydrochloride

  • MDL number MFCD16547588
  • Cas Number : 1240528-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63898
Availability details for
Quantity
  • Availability
  • Add to Cart