ENA295068159 Enamine

4-chloro-N-(propan-2-yl)aniline hydrochloride

  • MDL number MFCD16547590
  • Cas Number : 826-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63905
Availability details for
Quantity
  • Availability
  • Add to Cart