ENA295068169 Enamine

5-methyl-5-[3-(2-oxo-1,3-oxazolidin-3-yl)phenyl]imidazolidine-2,4-dione

  • MDL number MFCD16547602
  • Cas Number : 1240526-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63938
Availability details for
Quantity
  • Availability
  • Add to Cart