ENA295068170 Enamine

5-methyl-5-[4-(1H-1,2,3,4-tetrazol-1-yl)phenyl]imidazolidine-2,4-dione

  • MDL number MFCD16547603
  • Cas Number : 1240526-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63941
Availability details for
Quantity
  • Availability
  • Add to Cart