ENA295068178 Enamine

4-({[1-(ethanesulfonyl)piperidin-4-yl]carbamoyl}amino)benzoic acid

  • MDL number MFCD16547611
  • Cas Number : 1240529-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63994
Availability details for
Quantity
  • Availability
  • Add to Cart