ENA295068181 Enamine

2-amino-N-(4-bromo-2-fluorophenyl)-2-phenylacetamide hydrochloride

  • MDL number MFCD16547615
  • Cas Number : 1251922-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64033
Availability details for
Quantity
  • Availability
  • Add to Cart