ENA295068183 Enamine

N-(4-methylphenyl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16547617
  • Cas Number : 1251923-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64055
Availability details for
Quantity
  • Availability
  • Add to Cart