ENA295068189 Enamine

2-(4-methylphenyl)-1-(piperazin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD16547623
  • Cas Number : 1240527-10-9
  • Molecular Weight : 254.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(4-methylphenyl)acetyl]piperazine hydrochloride
Inchi Code 1S/C13H18N2O.ClH/c1-11-2-4-12(5-3-11)10-13(16)15-8-6-14-7-9-15;/h2-5,14H,6-10H2,1H3;1H
Inchi Key YBZNEBJHFZXFAB-UHFFFAOYSA-N

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64091
Availability details for
Quantity
  • Availability
  • Add to Cart