ENA295068197 Enamine

[1-(4-chlorophenyl)ethyl]hydrazine dihydrochloride

  • MDL number MFCD16547631
  • Cas Number : 1240528-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64113
Availability details for
Quantity
  • Availability
  • Add to Cart