ENA295068202 Enamine

methyl 1-(piperidin-4-yl)-1H-pyrrole-2-carboxylate hydrochloride

  • MDL number MFCD16547636
  • Cas Number : 1240528-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64141
Availability details for
Quantity
  • Availability
  • Add to Cart