ENA295068205 Enamine

2-(2,6-dichlorophenyl)propan-2-amine hydrochloride

  • MDL number MFCD16547639
  • Cas Number : 1240528-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236...238
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64172

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64172
Availability details for
Quantity
  • Availability
  • Add to Cart