ENA295068206 Enamine

methyl 1,1-dioxo-3,4-dihydro-2H-1lambda6,2,4-benzothiadiazine-3-carboxylate

  • MDL number MFCD16547640
  • Cas Number : 1251923-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64179
Availability details for
Quantity
  • Availability
  • Add to Cart