ENA295068209 Enamine

5-(1-benzofuran-2-yl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD16547643
  • Cas Number : 1240526-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64188
Availability details for
Quantity
  • Availability
  • Add to Cart