ENA295068212 Enamine

6-[(difluoromethyl)sulfanyl]-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD16547646
  • Cas Number : 1240528-46-4
  • Molecular Weight : 228.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64200
Availability details for
Quantity
  • Availability
  • Add to Cart