ENA295068213 Enamine

2-(trifluoromethane)sulfonylphenol

  • MDL number MFCD16547647
  • Cas Number : 32858-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64211

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64211
Availability details for
Quantity
  • Availability
  • Add to Cart