ENA295068221 Enamine

1-(piperidin-4-yl)-4,5,6,7-tetrahydro-1H-1,2,3-benzotriazole dihydrochloride

  • MDL number MFCD16547656
  • Cas Number : 1240528-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64271

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64271
Availability details for
Quantity
  • Availability
  • Add to Cart