ENA295068224 Enamine

3-chloro-2-[(difluoromethyl)sulfanyl]aniline hydrochloride

  • MDL number MFCD16547662
  • Cas Number : 1240527-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64336
Availability details for
Quantity
  • Availability
  • Add to Cart