ENA295068225 Enamine

5,6-dimethoxy-2,3-dihydro-1lambda6,2-benzothiazole-1,1,3-trione

  • MDL number MFCD16547663
  • Cas Number : 60987-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64340
Availability details for
Quantity
  • Availability
  • Add to Cart