ENA295068227 Enamine

3-bromo-4-chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD16547665
  • Cas Number : 1240526-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64396
Availability details for
Quantity
  • Availability
  • Add to Cart