ENA295068231 Enamine

8-ethyl-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD16547670
  • Cas Number : 191608-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64417
Availability details for
Quantity
  • Availability
  • Add to Cart