ENA295068246 Enamine

3-{2,3-dimethylpyrazolo[1,5-a]pyrimidin-6-yl}propan-1-ol

  • MDL number MFCD16547688
  • Cas Number : 1240526-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64504
Availability details for
Quantity
  • Availability
  • Add to Cart