ENA295068249 Enamine

N-[3-(chloromethyl)phenyl]acetamide

  • MDL number MFCD16547691
  • Cas Number : 90942-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64514
Availability details for
Quantity
  • Availability
  • Add to Cart