ENA295068252 Enamine

4-{[(3-methoxyphenyl)methyl]amino}cyclohexan-1-ol hydrochloride

  • MDL number MFCD16547694
  • Cas Number : 1240527-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64520
Availability details for
Quantity
  • Availability
  • Add to Cart