ENA295068258 Enamine

4-(pyridin-2-ylsulfanyl)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD16547701
  • Cas Number : 1240527-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64584
Availability details for
Quantity
  • Availability
  • Add to Cart