ENA295068259 Enamine

1-(piperazin-1-yl)-2-(pyridin-2-yl)ethan-1-one dihydrochloride

  • MDL number MFCD16547702
  • Cas Number : 1240527-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64585
Availability details for
Quantity
  • Availability
  • Add to Cart