ENA295068263 Enamine

2-(benzylamino)-2-[4-(trifluoromethyl)phenyl]acetic acid

  • MDL number MFCD16547706
  • Cas Number : 1251922-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64603
Availability details for
Quantity
  • Availability
  • Add to Cart