ENA295068268 Enamine

5-bromo-6-(bromomethyl)imidazo[2,1-b][1,3]thiazole hydrobromide

  • MDL number MFCD16547711
  • Cas Number : 135063-74-0
  • Molecular Weight : 376.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64653
Availability details for
Quantity
  • Availability
  • Add to Cart