ENA295068269 Enamine

2-(2-phenylethoxy)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD16547712
  • Cas Number : 1240526-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64658
Availability details for
Quantity
  • Availability
  • Add to Cart