ENA295068279 Enamine

(5-phenyl-1,2,4-oxadiazol-3-yl)methanediol

  • MDL number MFCD16547722
  • Cas Number : 73217-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64728
Availability details for
Quantity
  • Availability
  • Add to Cart