ENA295068281 Enamine

1-[(2-fluorophenyl)methyl]-1H-imidazol-2-amine hydrochloride

  • MDL number MFCD16547724
  • Cas Number : 1240527-63-2
  • Molecular Weight : 227.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64739
Availability details for
Quantity
  • Availability
  • Add to Cart