ENA295068282 Enamine

2-[3-(chlorosulfonyl)-2,6-dimethylphenoxy]acetic acid

  • MDL number MFCD16547725
  • Cas Number : 1240528-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64749
Availability details for
Quantity
  • Availability
  • Add to Cart