ENA295068284 Enamine

3-(2,5-dimethyl-1H-pyrrol-1-yl)propan-1-ol

  • MDL number MFCD16547727
  • Cas Number : 54609-15-3
  • Molecular Weight : 153.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64763
Availability details for
Quantity
  • Availability
  • Add to Cart