ENA295068287 Enamine

6-(furan-2-yl)-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD16547731
  • Cas Number : 1240527-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64802
Availability details for
Quantity
  • Availability
  • Add to Cart