ENA295068383 Enamine

5-(2,6-difluorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD14708194
  • Cas Number : 397844-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62832
Availability details for
Quantity
  • Availability
  • Add to Cart