ENA295068385 Enamine

2-(4,6-dimethyl-2-oxo-1,2-dihydropyrimidin-1-yl)ethanethioamide

  • MDL number MFCD09935129
  • Cas Number : 1016769-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62836
Availability details for
Quantity
  • Availability
  • Add to Cart