ENA295068386 Enamine

{1-[(3-aminophenyl)methyl]piperidin-4-yl}methanol

  • MDL number MFCD11132973
  • Cas Number : 1094353-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62840

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62840
Availability details for
Quantity
  • Availability
  • Add to Cart