ENA295068406 Enamine

2-(4,4-dimethyl-1,2,3,4-tetrahydronaphthalen-1-yl)acetic acid

  • MDL number MFCD11200492
  • Cas Number : 1038360-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63859
Availability details for
Quantity
  • Availability
  • Add to Cart