ENA295068408 Enamine

3-[(4-sulfamoylphenyl)amino]propanoic acid

  • MDL number MFCD00159140
  • Cas Number : 6541-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63862
Availability details for
Quantity
  • Availability
  • Add to Cart