ENA295068410 Enamine

1-(3-fluorophenyl)-2-(1H-imidazol-1-yl)ethan-1-one

  • MDL number MFCD12061374
  • Cas Number : 630403-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63864
Availability details for
Quantity
  • Availability
  • Add to Cart