ENA295068411 Enamine

1-(2-bromophenyl)-2-(1H-imidazol-1-yl)ethan-1-one

  • MDL number MFCD12061379
  • Cas Number : 763036-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63865
Availability details for
Quantity
  • Availability
  • Add to Cart